Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4''-Dinitro-p-terphenyl, 98+%

$173.28 - $697.64

Chemical Identifiers

Chemical Name or Material 4,4''-Dinitro-p-terphenyl
CAS 3282-11-9
Molecular Formula C18H12N2O4
Formula Weight 320.31
MDL Number MFCD00051743
Beilstein 2160043
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2205906

  SDS
alfa aesar™
B2205906
5g Each for $173.28

AAB2205914

  SDS
alfa aesar™
B2205914
25g Each for $697.64
Description & Specifications

Specifications

Formula Weight 320.31
MDL Number MFCD00051743
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula C18H12N2O4
Melting Point 276° to 280°C
Assay ≥98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Beilstein 2160043
Chemical Name or Material 4,4''-Dinitro-p-terphenyl
CAS 3282-11-9
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.