Please login to your online account to display your discounted pricing

4,4-Dimethylcyclohexan-1-ol, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTBG00026DA

 View more versions of this product

Catalog No. BTBG00026DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 128.21
Quantity 1g
Assay 95%
CAS Max % 100.0
Health Hazard 2 GHS P Statement Causes serious eye irritation. Causes skin irritation.
Molecular Formula C8H16O
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Chemical Name or Material 4, 4-Dimethylcyclohexan-1-ol
Assay Percent Range 95%
MDL Number MFCD00101954
CAS 932-01-4
CAS Min % 94.5
Packaging Amber Glass Bottle
Health Hazard 1 Warning

Specifications

Chemical Name or Material 4, 4-Dimethylcyclohexan-1-ol
CAS 932-01-4
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H16O
Formula Weight 128.21
MDL Number MFCD00101954
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

GHS P Statement Causes serious eye irritation. Causes skin irritation.