Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4-Dimethyl-3-thiosemicarbazide, 95%

Chemical Identifiers

Chemical Name or Material 4,4-Dimethyl-3-thiosemicarbazide
CAS 6926-58-5
Molecular Formula C3H9N3S
Formula Weight 129.18
MDL Number MFCD00041308
UN Number UN2811
Beilstein 1747186
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1033703

  SDS
alfa aesar™
L1033703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1033706

  SDS
alfa aesar™
L1033706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.18
Chemical Name or Material 4,4-Dimethyl-3-thiosemicarbazide
Molecular Formula C3H9N3S
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
MDL Number MFCD00041308
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 95%
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 142° to 147°C
Beilstein 1747186
Odor Odorless
UN Number UN2811
CAS 6926-58-5

Applications
4,4-Dimethyl-3-thiosemicarbazide may be employed as chemical additive to investigate the corrosion inhibition of mild steel in acidic conditions.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.