Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4-Dimethyl-2-phenyl-2-oxazoline, 97%

$68.42 - $189.85

Chemical Identifiers

Chemical Name or Material 4,4-Dimethyl-2-phenyl-2-oxazoline
CAS 19312-06-2
Molecular Formula C11H13NO
Formula Weight 175.23
MDL Number MFCD00030446
Beilstein 132225
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2505822

  SDS
alfa aesar™
B2505822
100g Each for $68.42

AAB2505836

  SDS
alfa aesar™
B2505836
500g Each for $189.85
Description & Specifications

Specifications

Formula Weight 175.23
Flash Point 102°C (215°F)
MDL Number MFCD00030446
CAS 19312-06-2
Molecular Formula C11H13NO
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Melting Point 20° to 24°C
Boiling Point 124°C (20mmHg)
Quantity 100g
Chemical Name or Material 4,4-Dimethyl-2-phenyl-2-oxazoline
Beilstein 132225
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.