Alfa Aesar™ 4,4-Dimethyl-2-cyclohexen-1-one, 97+%
Manufacturer:  Alfa Aesar™ 1716206
 View more versions of this product
Catalog No. AA1716206
Specifications
Specifications
4,4-Dimethyl-2-cyclohexen-1-one
≥97%
C8H12O
73°C to 74°C (14mmHg)
64° (148°F)
HAUNPYVLVAIUOO-UHFFFAOYSA-N
4,4-dimethylcyclohex-2-en-1-one
136839
Liquid
1073-13-8
0.944g/mL
MFCD00009695
5g
Air Sensitive
CC1(CCC(=O)C=C1)C
124.183
124.18
Chemical Identifiers
1073-13-8
124.183
HAUNPYVLVAIUOO-UHFFFAOYSA-N
4,4-dimethylcyclohex-2-en-1-one
C8H12O
MFCD00009695
136839
CC1(CCC(=O)C=C1)C
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents