missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Dimethyl-2,2'-bipyridyl 1-Oxide 98.0+%, TCI America™
$160.92 - $560.75
Chemical Identifiers
CAS | 81998-03-0 |
---|---|
Molecular Formula | C12H12N2O |
Molecular Weight (g/mol) | 200.241 |
MDL Number | MFCD11042487 |
InChI Key | VNSKKRAUVGCMPH-UHFFFAOYSA-N |
PubChem CID | 13586792 |
IUPAC Name | 4-methyl-2-(4-methylpyridin-2-yl)-1-oxidopyridin-1-ium |
SMILES | CC1=CC(=NC=C1)C2=[N+](C=CC(=C2)C)[O-] |
Chemical Identifiers
81998-03-0 | |
200.241 | |
VNSKKRAUVGCMPH-UHFFFAOYSA-N | |
4-methyl-2-(4-methylpyridin-2-yl)-1-oxidopyridin-1-ium |
C12H12N2O | |
MFCD11042487 | |
13586792 | |
CC1=CC(=NC=C1)C2=[N+](C=CC(=C2)C)[O-] |
Specifications
81998-03-0 | |
Yellow | |
MFCD11042487 | |
4,4′C-Dimethyl-2,2′C-bipyridine 1-Oxide | |
CC1=CC(=NC=C1)C2=[N+](C=CC(=C2)C)[O-] | |
200.241 | |
200.24 | |
Crystalline Powder |
87°C | |
C12H12N2O | |
1 g | |
VNSKKRAUVGCMPH-UHFFFAOYSA-N | |
4-methyl-2-(4-methylpyridin-2-yl)-1-oxidopyridin-1-ium | |
13586792 | |
≥98.0% (GC) | |
4,4′-Dimethyl-2,2′-bipyridyl 1-Oxide |
Safety and Handling
TSCA : No