Please login to your online account to display your discounted pricing

4,4-Dimethyl-1,3-cyclohexanedione 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4, 4-Dimethyl-1, 3-cyclohexanedione
CAS 562-46-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H12O2
Formula Weight 140.18
MDL Number MFCD00075224
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381260050

  SDS
thermo fisher scientific
AC381260050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC381260250

  SDS
thermo fisher scientific
AC381260250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Needles
Formula Weight 140.18
Quantity 5g
Color White
Melting Point 102° to 107°C
CAS Max % 100.0
Chemical Name or Material 4, 4-Dimethyl-1, 3-cyclohexanedione
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H12O2
MDL Number MFCD00075224
CAS Min % 97.5
CAS 562-46-9
Packaging Glass bottle