Learn More
4,4'-Dimethoxytrityl Chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 146021000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4, 4'-Dimethoxytrityl chloride | |
97.5 | |
121°C to 127°C | |
Authentic | |
Glass bottle | |
C6H5C(C6H4OCH3)2Cl | |
100 g | |
4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl | |
COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1 | |
338.83 | |
338.82 | |
Powder |
40615-36-9 | |
100.0 | |
Pink | |
97.5% min. (ex Cl) (Argentometry) | |
C21H19ClO2 | |
MFCD00008409 | |
01,188 | |
JBWYRBLDOOOJEU-UHFFFAOYSA-N | |
1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene | |
96831 | |
98% |
Chemical Identifiers
40615-36-9 | |
338.83 | |
JBWYRBLDOOOJEU-UHFFFAOYSA-N | |
96831 | |
COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1 |
C21H19ClO2 | |
MFCD00008409 | |
4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl | |
1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 255-002-6
RUO – Research Use Only