Please login to your online account to display your discounted pricing

4,4'-Dimethoxytrityl Chloride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  146020050

 View more versions of this product

Catalog No. AC146020050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ex Cl)
Formula Weight 338.82
Physical Form Powder
Quantity 5g
Color Pink
Fieser 01,188
Melting Point 121° to 127°C
CAS Max % 100.0
Synonym 4, 4'-Dimethoxytriphenylmethylechloride
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 40615-36-9
Chemical Name or Material 4, 4'-Dimethoxytrityl chloride
Assay Percent Range 98%
MDL Number MFCD00008409
Molecular Formula C21H19ClO2
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 97.5
Linear Formula C6H5C(C6H4OCH3)2Cl
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4, 4'-Dimethoxytrityl chloride
CAS 40615-36-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (ex Cl)
Assay Percent Range 98%
Molecular Formula C21H19ClO2
Linear Formula C6H5C(C6H4OCH3)2Cl
Formula Weight 338.82
MDL Number MFCD00008409
Synonym 4, 4'-Dimethoxytriphenylmethylechloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents