missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Dimethoxytrityl chloride, 98%

Manufacturer:  Alfa Aesar™ A1162606

 View more versions of this product

Catalog No. AAA1162606


 

Description

Description

Applications
4,4′-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.

Solubility
Soluble in chloroform and methanol. Partially soluble in water

Notes
Moisture sensitive, hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

4,4'-Dimethoxytrityl chloride
5g
98%
MFCD00008409
4,4'-Dimethoxytriphenylmethyl chloride; DMT
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)OC)Cl
338.831
338.83
121°C to 127°C
40615-36-9
C21H19ClO2
2471942
JBWYRBLDOOOJEU-UHFFFAOYSA-N
1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
96831

Chemical Identifiers

40615-36-9
338.831
JBWYRBLDOOOJEU-UHFFFAOYSA-N
96831
C21H19ClO2
MFCD00008409
4,4'-Dimethoxytriphenylmethyl chloride; DMT
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents