missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Dimethoxybenzophenone 97%, ACROS Organics™

Manufacturer:  Acros Organics AC161741000

 View more versions of this product

Catalog No. AC161741000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4,4′-Dimethoxybenzophenone
Plastic bottle
141.0°C to 146.0°C
97%
96.0
97%
(CH3OC6H4)2CO
08, 317
RFVHVYKVRGKLNK-UHFFFAOYSA-N
bis(4-methoxyphenyl)methanone
7032
Powder
Authentic
Beige to Yellow
100g
90-96-0
100.0
C15H14O3
MFCD00008404
4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
COC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)OC
242.274
242.27
97%

Chemical Identifiers

90-96-0
242.274
RFVHVYKVRGKLNK-UHFFFAOYSA-N
7032
COC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)OC
C15H14O3
MFCD00008404
4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
bis(4-methoxyphenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-028-0

TSCA : TSCA

SDS Documents