Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Diiodobiphenyl, 99%

Chemical Identifiers

Chemical Name or Material 4,4'-Diiodobiphenyl
CAS 3001-15-8
Molecular Formula C12H8I2
Formula Weight 406
MDL Number MFCD00001057
UN Number UN3152
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2197022

 
alfa aesar™
B2197022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2197014

 
alfa aesar™
B2197014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 406
Chemical Name or Material 4,4'-Diiodobiphenyl
TSCA TSCA
Melting Point 205° to 211°C
Assay 99%
Molecular Formula C12H8I2
Quantity 100g
MDL Number MFCD00001057
CAS 3001-15-8
UN Number UN3152
DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID

Applications
4,4′-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4′-diiodobipheny enhanced the expression of the luciferase gene.

Solubility
Soluble in hot Toluene(very faint turbidity).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.