Learn More
4,4'-Diiodobiphenyl, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2197022
Description
4,4?-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4?-diiodobipheny enhanced the expression of the luciferase gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4′-diiodobipheny enhanced the expression of the luciferase gene.
Solubility
Soluble in hot Toluene(very faint turbidity).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
4,4'-Diiodobiphenyl | |
205°C to 211°C | |
MFCD00001057 | |
UN3152 | |
4,4'-diiodobiphenyl, 4,4'-diiodo-1,1'-biphenyl, 4,4'-diiododiphenyl, 1,1'-biphenyl, 4,4'-diiodo, 1-iodo-4-4-iodophenyl benzene, 4,4-diiodobiphenyl, 4-4'-iodophenyl iodobenzene, 4,4;-diiodobiphenyl, 4,4'-diiododophenyl, pract, 4-iodo-1-4-iodophenyl benzene | |
GPYDMVZCPRONLW-UHFFFAOYSA-N | |
1-iodo-4-(4-iodophenyl)benzene | |
76348 | |
99% |
3001-15-8 | |
C12H8I2 | |
100 g | |
Light sensitive | |
Soluble in hot Toluene(very faint turbidity). | |
C1=CC(=CC=C1C2=CC=C(C=C2)I)I | |
406.005 | |
406 |
Chemical Identifiers
3001-15-8 | |
406.005 | |
GPYDMVZCPRONLW-UHFFFAOYSA-N | |
76348 | |
C1=CC(=CC=C1C2=CC=C(C=C2)I)I |
C12H8I2 | |
MFCD00001057 | |
4,4'-diiodobiphenyl, 4,4'-diiodo-1,1'-biphenyl, 4,4'-diiododiphenyl, 1,1'-biphenyl, 4,4'-diiodo, 1-iodo-4-4-iodophenyl benzene, 4,4-diiodobiphenyl, 4-4'-iodophenyl iodobenzene, 4,4;-diiodobiphenyl, 4,4'-diiododophenyl, pract, 4-iodo-1-4-iodophenyl benzene | |
1-iodo-4-(4-iodophenyl)benzene |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
EINECSNumber : 221-080-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only