Learn More
4,4'-Dihydroxydiphenylmethane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1141703
Description
4,4'-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.
Solubility
It is soluble in ethanol, ether, chloroform, alkali; slightly soluble in DMSO, insoluble in carbon disulfide and water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions.
Specifications
4,4'-Dihydroxydiphenylmethane | |
159°C to 163°C | |
MFCD00002385 | |
2049425 | |
It is soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO,insoluble in carbon disulfide and water. | |
C1=CC(=CC=C1CC2=CC=C(C=C2)O)O | |
200.237 | |
CHEBI:34575 | |
98% |
620-92-8 | |
C13H12O2 | |
1 g | |
4,4'-methylenediphenol, bis 4-hydroxyphenyl methane, 4,4'-dihydroxydiphenylmethane, phenol, 4,4'-methylenebis, p-p-hydroxybenzyl phenol, bis p-hydroxyphenyl methane, phenol, 4,4'-methylenedi, 4,4'-methylenebisphenol, p,p'-bis hydroxyphenyl methane, 4-4-hydroxyphenyl methyl phenol | |
PXKLMJQFEQBVLD-UHFFFAOYSA-N | |
4-[(4-hydroxyphenyl)methyl]phenol | |
12111 | |
200.24 |
Chemical Identifiers
620-92-8 | |
200.237 | |
PXKLMJQFEQBVLD-UHFFFAOYSA-N | |
12111 | |
4-[(4-hydroxyphenyl)methyl]phenol |
C13H12O2 | |
MFCD00002385 | |
4,4'-methylenediphenol, bis 4-hydroxyphenyl methane, 4,4'-dihydroxydiphenylmethane, phenol, 4,4'-methylenebis, p-p-hydroxybenzyl phenol, bis p-hydroxyphenyl methane, phenol, 4,4'-methylenedi, 4,4'-methylenebisphenol, p,p'-bis hydroxyphenyl methane, 4-4-hydroxyphenyl methyl phenol | |
CHEBI:34575 | |
C1=CC(=CC=C1CC2=CC=C(C=C2)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-658-2
RTECSNumber : SL9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only