Please login to your online account to display your discounted pricing

4,4'-Difluorodiphenylmethane, 99%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  243281000

 View more versions of this product

Catalog No. AC243281000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid After Melting
Formula Weight 204.22
Quantity 100mL
Color Clear colorless to yellow
EINECS Number 207-274-2
TSCA TSCA
Density 1.1500g/mL
Beilstein 05, IV, 1846
CAS Max % 100.0
Chemical Name or Material 4, 4'-Difluorodiphenylmethane
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C13 H10 F2
MDL Number MFCD00000360
CAS 457-68-1
Synonym 1, 1'-Methylenebis[4-fluorobenzene]
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5345 to 1.5365 (20°C, 589nm)

Specifications

Chemical Name or Material 4, 4'-Difluorodiphenylmethane
Name Note 99%
CAS 457-68-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13 H10 F2
Formula Weight 204.22
MDL Number MFCD00000360
Synonym 1, 1'-Methylenebis[4-fluorobenzene]
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents