Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4-Difluorocyclohexylamine hydrochloride, 98%

$200.41 - $610.24

Chemical Identifiers

Chemical Name or Material 4,4-Difluorocyclohexylamine hydrochloride
CAS 675112-70-6
Molecular Formula C6H11F2N·HCl
Formula Weight 171.62
MDL Number MFCD06246881
Synonym 1-Amino-4,4-difluorocyclohexane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25832MC

  SDS
alfa aesar™
H25832MC
100mg Each for $200.41

AAH25832ME

  SDS
alfa aesar™
H25832ME
500mg Each for $610.24
Description & Specifications

Specifications

Formula Weight 171.62
Melting Point ≈295°C (dec.)
Molecular Formula C6H11F2N·HCl
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Amino-4,4-difluorocyclohexane hydrochloride
Quantity 100mg
MDL Number MFCD06246881
Assay 98%
CAS 675112-70-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4,4-Difluorocyclohexylamine hydrochloride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.