missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Difluorobiphenyl, 99%, Thermo Scientific Chemicals
$121.38 - $121.38
Chemical Identifiers
CAS | 398-23-2 |
---|---|
Molecular Formula | C12H8F2 |
Molecular Weight (g/mol) | 190.193 |
MDL Number | MFCD00000349 |
InChI Key | PZDAAZQDQJGXSW-UHFFFAOYSA-N |
Synonym | 4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro |
PubChem CID | 9811 |
IUPAC Name | 1-fluoro-4-(4-fluorophenyl)benzene |
SMILES | C1=CC(=CC=C1C2=CC=C(C=C2)F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0836106
|
Thermo Scientific Chemicals
L0836106 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
398-23-2 | |
190.193 | |
PZDAAZQDQJGXSW-UHFFFAOYSA-N | |
9811 | |
C1=CC(=CC=C1C2=CC=C(C=C2)F)F |
C12H8F2 | |
MFCD00000349 | |
4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro | |
1-fluoro-4-(4-fluorophenyl)benzene |
Specifications
398-23-2 | |
254°C to 255°C | |
MFCD00000349 | |
UN3152 | |
14,3149 | |
PZDAAZQDQJGXSW-UHFFFAOYSA-N | |
1-fluoro-4-(4-fluorophenyl)benzene | |
9811 | |
99% |
87°C to 89°C | |
C12H8F2 | |
5 g | |
2044699 | |
4,4'-difluorobiphenyl, 4,4'-difluoro-1,1'-biphenyl, 4,4'-difluorodiphenyl, 1-fluoro-4-4-fluorophenyl benzene, unii-o0694gmk81, 1,1'-biphenyl, 4,4'-difluoro, 4-fluoro-1-4-fluorophenyl benzene, pubchem2016, 4,4'-difluorobipheny, biphenyl,4'-difluoro | |
C1=CC(=CC=C1C2=CC=C(C=C2)F)F | |
190.193 | |
190.19 | |
4,4'-Difluorobiphenyl |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
EINECSNumber : 206-910-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only