Please login to your online account to display your discounted pricing

4,4'-Difluorobenzophenone 99%, ACROS Organics™

$149.12 - $149.12

Chemical Identifiers

Chemical Name or Material 4, 4'-Difluorobenzophenone
Name Note 99%
CAS 345-92-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H8F2O
Linear Formula (FC6H4)2CO
Formula Weight 218.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151790050

  SDS
thermo fisher scientific
AC151790050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC151791000

  SDS
fisher scientific acros
151791000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC151790250

  SDS
thermo fisher scientific
151790250
Glass bottle 25g Each for $149.12
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 218.2
Quantity 5g
Color White to slightly yellow
EINECS Number 206-466-3
TSCA TSCA
RTECS Number DJ0685000
Melting Point 106° to 109°C
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 4, 4'-Difluorobenzophenone
Name Note 99%
Assay Percent Range 99%
Molecular Formula C13H8F2O
Linear Formula (FC6H4)2CO
MDL Number MFCD00000353
CAS 345-92-6
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Loss on Drying 0.25% max.
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.