Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Difluorobenzil, 99%

$30.45 - $458.05

Chemical Identifiers

Chemical Name or Material 4,4'-Difluorobenzil
CAS 579-39-5
Molecular Formula C14H8F2O2
Formula Weight 246.22
MDL Number MFCD00134541
Beilstein 1971736
Synonym 4,4'-Difluorodibenzoyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1155003

  SDS
alfa aesar™
A1155003
1g Each for $30.45

AAA1155006

  SDS
alfa aesar™
A1155006
5g Each for $102.31

AAA1155014

  SDS
alfa aesar™
A1155014
25g Each for $458.05
Description & Specifications

Specifications

Formula Weight 246.22
Chemical Name or Material 4,4'-Difluorobenzil
TSCA TSCA
Assay 99%
Molecular Formula C14H8F2O2
MDL Number MFCD00134541
CAS 579-39-5
Beilstein 1971736
Synonym 4,4'-Difluorodibenzoyl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 120° to 122°C
Quantity 1g

Applications
4,4′-Difluorobenzil is used in the preparation of 2,3-bis(4-fluorophenyl)thieno[3,4-b]pyrazine. It is also used in the synthesis of poly(ether-α-diketone)s, via nucleophilic substitution reaction with bisphenol A, 4,4′-difluoro-2,3-diphenylpyrazine (DPPF) and 4,4′-difluoro-5-methyl 2,3-diphenylpyrazine (MDPPF), via direct condensation with ethylenediamine and 1,2-diaminopropane, respectively, 2,3-bis(4-fluorophenyl)quinoxaline, via direct condensation with 1,2-phenylenediamine.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.