Learn More
4,4'-Difluorobenzhydryl chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1122609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,4'-Difluorobenzhydryl chloride | |
1.28 | |
>110°C (230°F) | |
1.557 | |
10 g | |
2052680 | |
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene | |
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F | |
238.662 | |
238.66 |
27064-94-4 | |
126°C to 128°C (1 mmHg) | |
C13H9ClF2 | |
MFCD00044329 | |
UN1760 | |
Moisture sensitive | |
FHPNLCLHMNPLEW-UHFFFAOYSA-N | |
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene | |
2723773 | |
98% |
Chemical Identifiers
27064-94-4 | |
238.662 | |
FHPNLCLHMNPLEW-UHFFFAOYSA-N | |
2723773 | |
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F |
C13H9ClF2 | |
MFCD00044329 | |
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene | |
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 248-201-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only