missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4-Diethylamino-2-butyn-1-ol, 98%

Manufacturer:  Alfa Aesar™ H5350114

 View more versions of this product

Catalog No. AAH5350114



Description

Description

Applications
4,4-Diethylamino-2-butyn-1-ol is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform and ethyl acetate. Soluble in water 11 g/L @25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

4,4-Diethylamino-2-butyn-1-ol
104°C (219°F)
231°C to 232°C
10575-25-4
(CH3CH2)2NCH2C≡CCH2OH
1-diethylaminobut-2-yn-4-ol
ACGZBRWTWOZSFU-UHFFFAOYSA-N
4-(diethylamino)but-2-yn-1-ol
82735
98%
0.952
1.479
25g
C8H15NO
MFCD00671356
Soluble in chloroform and ethyl acetate. Soluble in water 11g/L @25°C.
CCN(CC)CC#CCO
141.214
141.21

Chemical Identifiers

10575-25-4
141.214
ACGZBRWTWOZSFU-UHFFFAOYSA-N
82735
CCN(CC)CC#CCO
C8H15NO
MFCD00671356
1-diethylaminobut-2-yn-4-ol
4-(diethylamino)but-2-yn-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 234-163-6

TSCA : No

Recommended Storage : Ambient temperatures

Documents