missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Dichlorodiphenyl sulfone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 291182500

 View more versions of this product

Catalog No. AC291182500



Description

Description

Specifications

Specifications

4, 4'-Dichlorodiphenyl sulfone
Authentic
397.0°C
146.0°C to 151.0°C
97%
96.0
96% min. (GC)
MFCD00000619
1,1'-sulfonylbis 4-chlorobenzene, 4,4'-dichlorodiphenyl sulfone, 4,4'-dichlorodiphenylsulfone, 4-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, bis 4-chlorophenyl sulfone, bis p-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, p-chlorophenyl sulfone, sulfone, bis p-chlorophenyl
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
1-chloro-4-(4-chlorophenyl)sulfonylbenzene
6625
Fine Powder
233°C
Plastic bottle
White
250g
80-07-9
100.0
C12H8Cl2O2S
06, 327
Solubility in water: .. Other solubilities: soluble in alcohols,slightly soluble in aqueous solvents
C1=CC(=CC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)Cl
287.16
287.16
97%

Chemical Identifiers

80-07-9
287.16
GPAPPPVRLPGFEQ-UHFFFAOYSA-N
6625
C1=CC(=CC=C1S(=O)(=O)C2=CC=C(C=C2)Cl)Cl
C12H8Cl2O2S
MFCD00000619
1,1'-sulfonylbis 4-chlorobenzene, 4,4'-dichlorodiphenyl sulfone, 4,4'-dichlorodiphenylsulfone, 4-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, bis 4-chlorophenyl sulfone, bis p-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, p-chlorophenyl sulfone, sulfone, bis p-chlorophenyl
1-chloro-4-(4-chlorophenyl)sulfonylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 201-247-9

RTECSNumber : WR3450000

TSCA : TSCA

SDS Documents