Please login to your online account to display your discounted pricing

4,4'-Dichlorodiphenyl sulfone, 97%, ACROS Organics™


Quantity 25g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  291180250

 View more versions of this product

Catalog No. AC291180250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Fine Powder
Formula Weight 287.16
Quantity 25g
Color White
Beilstein 06, 327
Packaging Glass Bottle
RTECS Number WR3450000
TSCA TSCA
EINECS Number 201-247-9
Melting Point 146° to 151°C
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
MDL Number MFCD00000619
Infrared Spectrum Authentic
Synonym Bis(4-chlorophenyl) sulfone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Molecular Formula C12H8Cl2O2S
CAS 80-07-9
Chemical Name or Material 4, 4'-Dichlorodiphenyl sulfone
Assay Percent Range 97%

Specifications

Chemical Name or Material 4, 4'-Dichlorodiphenyl sulfone
Name Note 97%
CAS 80-07-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H8Cl2O2S
Formula Weight 287.16
MDL Number MFCD00000619
Synonym Bis(4-chlorophenyl) sulfone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents