missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Dichlorochalcone, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1071714

 View more versions of this product

Catalog No. AAL1071714



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,4'-Dichlorochalcone
156°C to 160°C
MFCD00018704
2331589
YMEMCRBNZSLQCQ-XCVCLJGOSA-N
(E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
5377011
≥98%
19672-59-4
C15H10Cl2O
25 g
4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one
ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1
277.14
277.15

Chemical Identifiers

19672-59-4
277.14
YMEMCRBNZSLQCQ-XCVCLJGOSA-N
5377011
ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1
C15H10Cl2O
MFCD00018704
4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one
(E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
Safety and Handling

Safety and Handling

RTECSNumber : UD5572635

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only