Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Dichlorobutyrophenone, 97%

$75.03 - $227.53

Chemical Identifiers

Chemical Name or Material 4,4'-Dichlorobutyrophenone
CAS 40877-09-6
Molecular Formula C10H10Cl2O
Formula Weight 217.09
MDL Number MFCD00018993
Beilstein 879782
Synonym 4-Chlorophenyl 3-chloropropyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2460814

  SDS
alfa aesar™
B2460814
25g Each for $75.03

AAB2460818

  SDS
alfa aesar™
B2460818
50g Each for $128.69

AAB2460822

  SDS
alfa aesar™
B2460822
100g Each for $227.53
Description & Specifications

Specifications

Formula Weight 217.09
Melting Point 28° to 32°C
MDL Number MFCD00018993
Quantity 25g
Beilstein 879782
Synonym 4-Chlorophenyl 3-chloropropyl ketone
Chemical Name or Material 4,4'-Dichlorobutyrophenone
CAS 40877-09-6
Assay 97%
Molecular Formula C10H10Cl2O