Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Dichlorobenzhydrol, 98%

$64.80 - $216.97

Chemical Identifiers

Chemical Name or Material 4,4'-Dichlorobenzhydrol
CAS 90-97-1
Molecular Formula C13H10Cl2O
Formula Weight 253.13
MDL Number MFCD00000629
Beilstein 1878717
Synonym Bis(4-chlorophenyl) carbinol; 4,4'-Dichlorodiphenylmethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0232714

  SDS
alfa aesar™
L0232714
25g Each for $64.80

AAL0232722

  SDS
alfa aesar™
L0232722
100g Each for $216.97
Description & Specifications

Specifications

Formula Weight 253.13
CAS 90-97-1
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 91° to 93°C
Quantity 25g
Chemical Name or Material 4,4'-Dichlorobenzhydrol
Assay 98%
Molecular Formula C13H10Cl2O
MDL Number MFCD00000629
Beilstein 1878717
Synonym Bis(4-chlorophenyl) carbinol; 4,4'-Dichlorodiphenylmethanol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
4,4′-Dichlorobenzhydrol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.