missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Dibromo-1,1′-spirobiindane-7,7′-diol 98.0+%, TCI America™

$339.56 - $1,177.89

Chemical Identifiers

CAS 681481-91-4
Molecular Formula C17H14Br2O2
Molecular Weight (g/mol) 410.105
MDL Number MFCD16660947
InChI Key OZVMVFKUNSNXQC-UHFFFAOYSA-N
Synonym 4,4′C-Dibromo-7,7′C-dihydroxy-1,1′C-spirobiindane
PubChem CID 11235237
IUPAC Name 7,7'-dibromo-3,3'-spirobi[1,2-dihydroindene]-4,4'-diol
SMILES C1CC2(CCC3=C(C=CC(=C32)O)Br)C4=C(C=CC(=C41)Br)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D43321G
View Documents
Tci America
D43321G
1g Each for $339.56
 
D43325G
View Documents
Tci America
D43325G
5g Each for $1,177.89
 
Specifications

Chemical Identifiers

681481-91-4
410.105
OZVMVFKUNSNXQC-UHFFFAOYSA-N
11235237
C1CC2(CCC3=C(C=CC(=C32)O)Br)C4=C(C=CC(=C41)Br)O
C17H14Br2O2
MFCD16660947
4,4′C-Dibromo-7,7′C-dihydroxy-1,1′C-spirobiindane
7,7'-dibromo-3,3'-spirobi[1,2-dihydroindene]-4,4'-diol

Specifications

Undesignated
1g
C17H14Br2O2
4,4′C-Dibromo-7,7′C-dihydroxy-1,1′C-spirobiindane
C1CC2(CCC3=C(C=CC(=C32)O)Br)C4=C(C=CC(=C41)Br)O
410.105
410.11
≥98.0% (GC)
148°C
681481-91-4
MFCD16660947
OZVMVFKUNSNXQC-UHFFFAOYSA-N
7,7'-dibromo-3,3'-spirobi[1,2-dihydroindene]-4,4'-diol
11235237
Crystal-Powder at 20°C
4,4′-Dibromo-1,1′-spirobiindane-7,7′-diol
Safety and Handling

Safety and Handling

TSCA : No