Please login to your online account to display your discounted pricing

4,4'-diaminobenzanilide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4, 4'-Diaminobenzanilide
CAS 785-30-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13 H13 N3 O
Linear Formula NH2C6H4C(O)NHC6H4NH2
Formula Weight 227.27
MDL Number MFCD00025361
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377031000

  SDS
thermo fisher scientific
AC377031000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 227.27
Quantity 100g
Color Off-white to gray-beige
Melting Point 203° to 207°C
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
CAS 785-30-8
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00025361
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
CAS Min % 97.5
Chemical Name or Material 4, 4'-Diaminobenzanilide
Infrared Spectrum Authentic
Linear Formula NH2C6H4C(O)NHC6H4NH2
Molecular Formula C13 H13 N3 O
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.