Learn More
4,4'-Diaminobenzanilide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2241909
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,4'-Diaminobenzanilide | |
200°C to 202°C | |
MFCD00025361 | |
2215469 | |
XPAQFJJCWGSXGJ-UHFFFAOYSA-N | |
4-amino-N-(4-aminophenyl)benzamide | |
69917 | |
98% |
785-30-8 | |
C13H13N3O | |
10 g | |
4,4'-diaminobenzanilide, 4-amino-n-4-aminophenyl benzamide, benzamide, 4-amino-n-4-aminophenyl, 4-aminobenzoyl-4'-aminoanilide, p,p'-diaminobenzanilide, 4,4'-diamino benzanilide, unii-00x4jm89uh, benzanilide, 4,4'-diamino, 4-aminophenyl-n-4-aminophenyl carboxamide, ccris 8916 | |
NC1=CC=C(NC(=O)C2=CC=C(N)C=C2)C=C1 | |
227.27 | |
227.27 |
Chemical Identifiers
785-30-8 | |
227.27 | |
XPAQFJJCWGSXGJ-UHFFFAOYSA-N | |
69917 | |
NC1=CC=C(NC(=O)C2=CC=C(N)C=C2)C=C1 |
C13H13N3O | |
MFCD00025361 | |
4,4'-diaminobenzanilide, 4-amino-n-4-aminophenyl benzamide, benzamide, 4-amino-n-4-aminophenyl, 4-aminobenzoyl-4'-aminoanilide, p,p'-diaminobenzanilide, 4,4'-diamino benzanilide, unii-00x4jm89uh, benzanilide, 4,4'-diamino, 4-aminophenyl-n-4-aminophenyl carboxamide, ccris 8916 | |
4-amino-N-(4-aminophenyl)benzamide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 212-321-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only