Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Diacetoxybiphenyl, 97%

$33.59 - $441.49

Chemical Identifiers

Chemical Name or Material 4,4'-Diacetoxybiphenyl
CAS 32604-29-8
Molecular Formula C16H14O4
Formula Weight 270.28
MDL Number MFCD00035973
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2140303

  SDS
alfa aesar™
B2140303
1g Each for $33.59

AAB2140306

  SDS
alfa aesar™
B2140306
5g Each for $116.03

AAB2140314

  SDS
alfa aesar™
B2140314
25g Each for $441.49
Description & Specifications

Specifications

Formula Weight 270.28
Melting Point 162° to 165°C
Molecular Formula C16H14O4
CAS 32604-29-8
Assay 97%
Quantity 1g
Chemical Name or Material 4,4'-Diacetoxybiphenyl
MDL Number MFCD00035973