Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Di-tert-butyldiphenylmethane, 99%

$86.19 - $1,118.01

Chemical Identifiers

Chemical Name or Material 4,4'-Di-tert-butyldiphenylmethane
CAS 19099-48-0
Molecular Formula C21H28
Formula Weight 280.45
MDL Number MFCD00043733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2182009

  SDS
alfa aesar™
B2182009
10g Each for $86.19

AAB2182018

  SDS
alfa aesar™
B2182018
50g Each for $302.86

AAB2182030

  SDS
alfa aesar™
B2182030
250g Each for $1,118.01
Description & Specifications

Specifications

Formula Weight 280.45
Quantity 10g
Chemical Name or Material 4,4'-Di-tert-butyldiphenylmethane
Melting Point 71° to 73°C
Assay 99%
MDL Number MFCD00043733
Molecular Formula C21H28
CAS 19099-48-0

Applications
It is employed as a intermediate for pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.