missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4'-Di-tert-butyl-2,2'-bipyridyl 98.0+%, TCI America™
Supplier: TCI America D31341G
Specifications
4,4′-Di-tert-butyl-2,2′-bipyridyl | |
161°C | |
C18H24N2 | |
1 g | |
TXNLQUKVUJITMX-UHFFFAOYSA-N | |
4-tert-butyl-2-(4-tert-butylpyridin-2-yl)pyridine | |
4249187 | |
≥98.0% (GC) |
72914-19-3 | |
White | |
MFCD01863731 | |
4,4′C-Di-tert-butyl-2,2′C-bipyridine | |
CC(C)(C)C1=CC(=NC=C1)C2=NC=CC(=C2)C(C)(C)C | |
268.404 | |
268.40 | |
Crystalline Powder |
Chemical Identifiers
72914-19-3 | |
268.404 | |
TXNLQUKVUJITMX-UHFFFAOYSA-N | |
4-tert-butyl-2-(4-tert-butylpyridin-2-yl)pyridine |
C18H24N2 | |
MFCD01863731 | |
4249187 | |
CC(C)(C)C1=CC(=NC=C1)C2=NC=CC(=C2)C(C)(C)C |
Safety and Handling
TSCA : No