Learn More
4,4'-Di-n-nonyl-2,2'-bipyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3147606
Description
Used in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the preparation of an artificial muscle with lamellar structure based on a nematic triblock copolymer. Also use as complex in cyclic voltammetric studies of copper complexes catalyzing atom transfer radical polymerization.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
4,4'-Di-n-nonyl-2,2'-bipyridine | |
61°C to 63°C | |
MFCD00800915 | |
4,4'-dinonyl-2,2'-bipyridine, 4,4'-dinonyl-2,2'-bipyridyl, 4-nonyl-2-4-nonylpyridin-2-yl pyridine, 2,2'-bipyridine, 4,4'-dinonyl, 4,4'-bis nonyl-2,2'-bipyridine, dnbpy, acmc-1cctu, bpdn01, 4,4-dinonyl-2,2-bipyridine, 4,4-dinoyl-2,2-dipyridyl | |
CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC | |
408.674 | |
408.67 |
142646-58-0 | |
C28H44N2 | |
5 g | |
VHJFWJXYEWHCGD-UHFFFAOYSA-N | |
4-nonyl-2-(4-nonylpyridin-2-yl)pyridine | |
4285174 | |
97% |
Chemical Identifiers
142646-58-0 | |
408.674 | |
VHJFWJXYEWHCGD-UHFFFAOYSA-N | |
4285174 | |
CCCCCCCCCC1=CC(=NC=C1)C2=NC=CC(=C2)CCCCCCCCC |
C28H44N2 | |
MFCD00800915 | |
4,4'-dinonyl-2,2'-bipyridine, 4,4'-dinonyl-2,2'-bipyridyl, 4-nonyl-2-4-nonylpyridin-2-yl pyridine, 2,2'-bipyridine, 4,4'-dinonyl, 4,4'-bis nonyl-2,2'-bipyridine, dnbpy, acmc-1cctu, bpdn01, 4,4-dinonyl-2,2-bipyridine, 4,4-dinoyl-2,2-dipyridyl | |
4-nonyl-2-(4-nonylpyridin-2-yl)pyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only