missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Cyanophenyl)-2-methylthiazole, 97%

Manufacturer:  Alfa Aesar™ H54478MD

 View more versions of this product

Catalog No. AAH54478MD



Specifications

Specifications

4-(4-Cyanophenyl)-2-methylthiazole
127406-79-5
MFCD01765038
FOKBMJMDQCPNJI-UHFFFAOYSA-N
4-(2-methyl-1,3-thiazol-4-yl)benzonitrile
716353
97%
250mg
C11H8N2S
4-2-methyl-1,3-thiazol-4-yl benzenecarbonitrile
CC1=NC(=CS1)C2=CC=C(C=C2)C#N
200.259
200.26

Chemical Identifiers

127406-79-5
200.259
FOKBMJMDQCPNJI-UHFFFAOYSA-N
716353
CC1=NC(=CS1)C2=CC=C(C=C2)C#N
C11H8N2S
MFCD01765038
4-2-methyl-1,3-thiazol-4-yl benzenecarbonitrile
4-(2-methyl-1,3-thiazol-4-yl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents