Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-cis-Stilbenedicarboxylic acid, 95%

$83.58 - $83.58

Chemical Identifiers

Chemical Name or Material 4,4'-cis-Stilbenedicarboxylic acid
CAS 133005-88-6
Molecular Formula C16H12O4
Formula Weight 268.27
MDL Number MFCD00013994
Beilstein 1978660
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0436403

  SDS
alfa aesar™
L0436403
1g Each for $83.58
Description & Specifications

Specifications

Formula Weight 268.27
Chemical Name or Material 4,4'-cis-Stilbenedicarboxylic acid
CAS 133005-88-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Molecular Formula C16H12O4
MDL Number MFCD00013994
Health Hazard 1 Warning
Melting Point >300°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1978660
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.