Please login to your online account to display your discounted pricing

4-[(4-chlorophenyl)thio]thiophene-3-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-[(4-Chlorophenyl)thio]thiophene-3-carboxylic acid
CAS 149609-86-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7ClO2S2
Formula Weight 270.76
MDL Number MFCD00052149
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM04905CB

 
fisher scientific company
KM04905CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KM04905DA

 
fisher scientific company
KM04905DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 270.76
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C11H7ClO2S2
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00052149
CAS 149609-86-9
Chemical Name or Material 4-[(4-Chlorophenyl)thio]thiophene-3-carboxylic acid