Please login to your online account to display your discounted pricing

4-[(4-chlorophenyl)thio]thiophene-3-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  KM04905CB

 View more versions of this product

Catalog No. KM04905CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 270.76
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C11H7ClO2S2
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00052149
CAS 149609-86-9
Chemical Name or Material 4-[(4-Chlorophenyl)thio]thiophene-3-carboxylic acid

Specifications

Chemical Name or Material 4-[(4-Chlorophenyl)thio]thiophene-3-carboxylic acid
CAS 149609-86-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7ClO2S2
Formula Weight 270.76
MDL Number MFCD00052149