Please login to your online account to display your discounted pricing

4-[(4-chlorophenyl)sulfonyl]-3-thiophenecarboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-[(4-chlorophenyl)sulfonyl]-3-thiophenecarboxylic acid
CAS 423769-77-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7ClO4S2
Formula Weight 302.76
MDL Number MFCD03407362
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00552CB

 
fisher scientific company
MO00552CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO00552DA

 
fisher scientific company
MO00552DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 302.76
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-[(4-chlorophenyl)sulfonyl]-3-thiophenecarboxylic acid
Packaging Amber Glass Bottle
Molecular Formula C11H7ClO4S2
MDL Number MFCD03407362
CAS 423769-77-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.