Please login to your online account to display your discounted pricing

4-[(4-chlorophenyl)sulfonyl]-3-thiophenecarboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00552DA

 View more versions of this product

Catalog No. MO00552DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 302.76
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-[(4-chlorophenyl)sulfonyl]-3-thiophenecarboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H7ClO4S2
MDL Number MFCD03407362
CAS 423769-77-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-[(4-chlorophenyl)sulfonyl]-3-thiophenecarboxylic acid
CAS 423769-77-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7ClO4S2
Formula Weight 302.76
MDL Number MFCD03407362
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents