Please login to your online account to display your discounted pricing

4-(4-Chlorophenyl)cyclohexanecarboxylic acid, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB02037DA

 View more versions of this product

Catalog No. SB02037DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 238.71
Quantity 1g
Assay 97%
CAS Min % 97.0
Linear Formula ClC6H4C6H10CO2H
Chemical Name or Material (E)-4-(4-Chlorophenyl)cyclohexanecarboxylic acid
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C13H15ClO2
MDL Number MFCD06797626
CAS 95233-37-7
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (E)-4-(4-Chlorophenyl)cyclohexanecarboxylic acid
CAS 95233-37-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H15ClO2
Linear Formula ClC6H4C6H10CO2H
Formula Weight 238.71
MDL Number MFCD06797626
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.