missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorophenyl)-4-hydroxypiperidine, 97%

Manufacturer:  Alfa Aesar™ A1213703

 View more versions of this product

Catalog No. AAA1213703


 

Specifications

Specifications

4- (4-Chlorophenyl)-4-hydroxypiperidine
1g
97%
MFCD00006001
4-(4-Chlorophenyl)-4-piperidinol
C1CNCCC1(C2=CC=C(C=C2)Cl)O
211.689
211.69
137°C to 141°C
39512-49-7
C11H14ClNO
165923
LZAYOZUFUAMFLD-UHFFFAOYSA-N
4-(4-chlorophenyl)piperidin-4-ol
38282

Chemical Identifiers

39512-49-7
211.689
LZAYOZUFUAMFLD-UHFFFAOYSA-N
38282
C11H14ClNO
MFCD00006001
4-(4-Chlorophenyl)-4-piperidinol
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents