Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorophenyl)-4-hydroxypiperidine, 97%

$38.62 - $38.62

Chemical Identifiers

Chemical Name or Material 4- (4-Chlorophenyl)-4-hydroxypiperidine
CAS 39512-49-7
Molecular Formula C11H14ClNO
Formula Weight 211.69
MDL Number MFCD00006001
Beilstein 165923
Synonym 4-(4-Chlorophenyl)-4-piperidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1213703

 
alfa aesar™
A1213703
1g Each for $38.62
Description & Specifications

Specifications

Formula Weight 211.69
Chemical Name or Material 4- (4-Chlorophenyl)-4-hydroxypiperidine
Assay 97%
Molecular Formula C11H14ClNO
MDL Number MFCD00006001
CAS 39512-49-7
Beilstein 165923
Synonym 4-(4-Chlorophenyl)-4-piperidinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 137° to 141°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.