Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorophenyl)-3-thiosemicarbazide, 97%

Chemical Identifiers

Chemical Name or Material 4-(4-Chlorophenyl)-3-thiosemicarbazide
CAS 22814-92-2
Molecular Formula C7H8ClN3S
Formula Weight 201.68
MDL Number MFCD00041295
UN Number UN2811
Beilstein 1818693
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1265703

  SDS
alfa aesar™
L1265703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1265706

  SDS
alfa aesar™
L1265706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.68
Chemical Name or Material 4-(4-Chlorophenyl)-3-thiosemicarbazide
Assay 97%
Molecular Formula C7H8ClN3S
MDL Number MFCD00041295
CAS 22814-92-2
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Odor Odorless
UN Number UN2811
Beilstein 1818693
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point ≈180°C
Quantity 1g

Applications
4-(4-Chlorophenyl)-3-thiosemicarbazide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.