missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorophenyl)-3-thiosemicarbazide, 97%

Manufacturer:  Alfa Aesar™ L1265703

 View more versions of this product

Catalog No. AAL1265703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Applications
4-(4-Chlorophenyl)-3-thiosemicarbazide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-(4-Chlorophenyl)-3-thiosemicarbazide
~180°C
22814-92-2
MFCD00041295
1818693
GFTWJLUVFRTLIL-UHFFFAOYSA-N
1-amino-3-(4-chlorophenyl)thiourea
706988
97%
Odorless
1g
C7H8ClN3S
UN2811
4-4-chlorophenyl-3-thiosemicarbazide, n-4-chlorophenyl hydrazinecarbothioamide, 3-amino-1-4-chlorophenyl thiourea, 1-amino-3-4-chlorophenyl thiourea, 4-chlorophenyl amino hydrazinomethane-1-thione, acmc-20amw7, maybridge1_006895, 4-p-chlorphenylthiosemicarbazid, 4-4-chlorophenyl thiosemicarbazide, 1-azanyl-3-4-chlorophenyl thiourea
C1=CC(=CC=C1NC(=S)NN)Cl
201.672
201.68

Chemical Identifiers

22814-92-2
201.672
GFTWJLUVFRTLIL-UHFFFAOYSA-N
706988
C1=CC(=CC=C1NC(=S)NN)Cl
C7H8ClN3S
MFCD00041295
4-4-chlorophenyl-3-thiosemicarbazide, n-4-chlorophenyl hydrazinecarbothioamide, 3-amino-1-4-chlorophenyl thiourea, 1-amino-3-4-chlorophenyl thiourea, 4-chlorophenyl amino hydrazinomethane-1-thione, acmc-20amw7, maybridge1_006895, 4-p-chlorphenylthiosemicarbazid, 4-4-chlorophenyl thiosemicarbazide, 1-azanyl-3-4-chlorophenyl thiourea
1-amino-3-(4-chlorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents