Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorophenoxy)benzaldehyde, 98%

$99.74 - $347.88

Chemical Identifiers

Chemical Name or Material 4-(4-Chlorophenoxy)benzaldehyde
CAS 61343-99-5
Molecular Formula C13H9ClO2
Formula Weight 232.67
MDL Number MFCD01631895
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5045103

  SDS
alfa aesar™
H5045103
1g Each for $347.88

AAH50451MD

  SDS
alfa aesar™
H50451MD
250mg Each for $99.74
Description & Specifications

Specifications

Formula Weight 232.67
CAS 61343-99-5
Chemical Name or Material 4-(4-Chlorophenoxy)benzaldehyde
Assay 98%
Molecular Formula C13H9ClO2
MDL Number MFCD01631895
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.