Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorobenzyl)pyridine, 97%

$59.38 - $59.38

Chemical Identifiers

Chemical Name or Material 4- (4-Chlorobenzyl)pyridine
CAS 4409-11-4
Molecular Formula C12H10ClN
Formula Weight 203.67
MDL Number MFCD00006444
Beilstein 1367705
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1067322

  SDS
alfa aesar™
A1067322
100g Each for $59.38

AAA1067336

  SDS
alfa aesar™
A1067336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4- (4-Chlorobenzyl)pyridine
Flash Point >110°C (230°F)
Boiling Point 123° to 125°C (6mmHg)
Quantity 100g
CAS 4409-11-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
RTECS Number US7204100
Assay 97%
Molecular Formula C12H10ClN
Formula Weight 203.67
MDL Number MFCD00006444
Beilstein 1367705
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.