Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chlorobenzoyl)piperidine, 97%

Chemical Identifiers

Chemical Name or Material 4-(4-Chlorobenzoyl)piperidine
CAS 53220-41-0
Formula Weight 223.7
MDL Number MFCD04113538
Beilstein 158791
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6474003

  SDS
alfa aesar™
H6474003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6474006

  SDS
alfa aesar™
H6474006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64740MD

  SDS
alfa aesar™
H64740MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 60° to 63°C
Formula Weight 223.7
Color Yellow
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
CAS 53220-41-0
Health Hazard 1 Warning
Beilstein 158791
Physical Form Solid
Chemical Name or Material 4-(4-Chlorobenzoyl)piperidine
MDL Number MFCD04113538
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.