missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 235120500

 View more versions of this product

Catalog No. AC235120500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
Glass bottle
135°C to 140°C
97%
96.0
97%
MFCD00066999
4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, piperidinalkohol, piperidine alcohol
C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O
279.69
279.69
97%
Authentic
Beige
50g
21928-50-7
100.0
C12H13ClF3NO
21, V,2, 409
RALRVIPTUXSBPO-UHFFFAOYSA-N
4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
89107
Fine Crystalline Powder

Chemical Identifiers

21928-50-7
279.69
RALRVIPTUXSBPO-UHFFFAOYSA-N
89107
C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O
C12H13ClF3NO
MFCD00066999
4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, piperidinalkohol, piperidine alcohol
4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 244-665-7

SDS Documents