missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%, ACROS Organics™

$143.80 - $143.80

Chemical Identifiers

CAS 21928-50-7
Molecular Formula C12H13ClF3NO
Molecular Weight (g/mol) 279.687
MDL Number MFCD00066999
InChI Key RALRVIPTUXSBPO-UHFFFAOYSA-N
Synonym 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol
PubChem CID 89107
IUPAC Name 4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
SMILES C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC235120050 SDS View Documents Acros Organics
235120050
5g Glass bottle Each for $143.80
AC235120500 SDS View Documents Acros Organics
235120500
50g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

21928-50-7
279.687
RALRVIPTUXSBPO-UHFFFAOYSA-N
89107
C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O
C12H13ClF3NO
MFCD00066999
4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol
4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol

Specifications

Authentic
Beige
5g
96.0
97%
MFCD00066999
4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl piperidin-4-ol, 4-4-chloro-3-trifluoromethyl phenyl-4-piperidinol, 4-piperidinol, 4-4-chloro-3-trifluoromethyl phenyl, 4-4-chloro-3-trifluoromethyl phenyl-4-hydroxypiperidine, piperidine alcohol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, 4-4-chloro-3-trifluoromethylphenyl-4-piperidinol, piperidinalkohol
C1CNCCC1(C2=CC(=C(C=C2)Cl)C(F)(F)F)O
279.687
279.69
≥96.0%
Glass bottle
135°C to 140°C
21928-50-7
100.0
C12H13ClF3NO
21, V,2, 409
RALRVIPTUXSBPO-UHFFFAOYSA-N
4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
89107
Fine Crystalline Powder
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 244-665-7