Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Chloro-2-nitrophenyl)morpholine, 97%

Chemical Identifiers

Chemical Name or Material 4-(4-Chloro-2-nitrophenyl)morpholine
CAS 65976-60-5
Formula Weight 242.66
Beilstein 218646
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6305703

 
alfa aesar™
H6305703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6305706

 
alfa aesar™
H6305706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63057MD

 
alfa aesar™
H63057MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.66
Chemical Name or Material 4-(4-Chloro-2-nitrophenyl)morpholine
Physical Form Solid
Assay 97%
CAS 65976-60-5
Beilstein 218646
Quantity 1g